Search
Search

FERMT2| Gene Symbol: FERMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561272.2 | XP_005561329.1 | 9 - 10 | 1378 | 86 | |
| XM_005561276.2 | XP_005561333.1 | 8 - 9 | 1345 | 86 | ||
| XM_005561277.2 | XP_005561334.1 | 8 - 9 | 1345 | 86 | ||
| XM_015453794.1 | XP_015309280.1 | 9 - 10 | 1378 | 86 | ||
| XM_015453795.1 | XP_015309281.1 | 8 - 9 | 1345 | 86 | ||
| XM_015453796.1 | XP_015309282.1 | 9 - 10 | 1378 | 86 | ||
Set Membership: |
|