Search
Search

STXBP2| Gene Symbol: STXBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587771.2 | XP_005587828.1 | 13 - 14 | 1179 | 88 | |
| XM_015440655.1 | XP_015296141.1 | 13 - 14 | 1979 | 88 | ||
Set Membership: |
|