Search
Search

WASF2| Gene Symbol: WASF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544274.2 | XP_005544331.1 | 6 - 7 | 910 | 108 | |
| XM_005544276.2 | XP_005544333.1 | 6 - 7 | 2182 | 108 | ||
| XM_015439968.1 | XP_015295454.1 | 7 - 8 | 1001 | 108 | ||
Set Membership: |
|