Search
Search

DGAT1| Gene Symbol: DGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564342.2 | XP_005564399.1 | 4 - 5 | 694 | 83 | |
| XM_005564343.2 | XP_005564400.1 | 4 - 5 | 688 | 83 | ||
| XM_005564344.2 | XP_005564401.1 | 4 - 5 | 688 | 83 | ||
Set Membership: |
|