Search
Search

C14H11orf54| Gene Symbol: C14H11orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579364.1 | XP_005579421.1 | 2 - 3 | 264 | 92 | |
| XM_005579365.2 | XP_005579422.1 | 2 - 3 | 819 | 92 | ||
| XM_005579366.2 | XP_005579423.1 | 2 - 3 | 309 | 92 | ||
Set Membership: |
|