Search
Search

AKAP8L| Gene Symbol: AKAP8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588278.2 | XP_005588335.1 | 10 - 11 | 1238 | 69 | |
| XM_005588279.2 | XP_005588336.1 | 9 - 10 | 1356 | 69 | ||
Set Membership: |
|