Search
Search

TTLL4| Gene Symbol: TTLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574310.2 | XP_005574367.1 | 18 - 19 | 3653 | 104 | |
| XM_005574311.2 | XP_005574368.1 | 19 - 20 | 3774 | 104 | ||
| XM_015432798.1 | XP_015288284.1 | 20 - 21 | 3821 | 104 | ||
| XM_015432799.1 | XP_015288285.1 | 20 - 21 | 3817 | 104 | ||
| XM_015432800.1 | XP_015288286.1 | 19 - 20 | 3726 | 104 | ||
| XM_015432803.1 | XP_015288289.1 | 20 - 21 | 3504 | 104 | ||
Set Membership: |
|