Search
Search

ARHGEF17| Gene Symbol: ARHGEF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579032.2 | XP_005579089.1 | 9 - 10 | 4518 | 68 | |
| XM_005579033.2 | XP_005579090.1 | 9 - 10 | 4515 | 68 | ||
| XM_005579034.2 | XP_005579091.1 | 9 - 10 | 4519 | 68 | ||
Set Membership: |
|