Search
Search

FOXJ3| Gene Symbol: FOXJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543752.2 | XP_005543809.1 | 12 - 13 | 1979 | 62 | |
| XM_005543753.1 | XP_005543810.1 | 12 - 13 | 2019 | 62 | ||
| XM_005543756.2 | XP_005543813.1 | 11 - 12 | 1826 | 62 | ||
| XM_015437555.1 | XP_015293041.1 | 12 - 13 | 1960 | 62 | ||
| XM_015437559.1 | XP_015293045.1 | 12 - 13 | 2084 | 62 | ||
| XM_015437564.1 | XP_015293050.1 | 12 - 13 | 2040 | 62 | ||
| XM_015437567.1 | XP_015293053.1 | 11 - 12 | 1895 | 62 | ||
Set Membership: |
|