Search
Search

CFAP61| Gene Symbol: CFAP61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568227.2 | XP_005568284.1 | 22 - 23 | 2936 | 147 | |
| XM_015457741.1 | XP_015313227.1 | 22 - 23 | 2951 | 147 | ||
| XM_015457742.1 | XP_015313228.1 | 22 - 23 | 2896 | 147 | ||
| XM_015457743.1 | XP_015313229.1 | 22 - 23 | 2905 | 147 | ||
| XM_015457744.1 | XP_015313230.1 | 22 - 23 | 2951 | 147 | ||
| XM_015457745.1 | XP_015313231.1 | 22 - 23 | 2951 | 147 | ||
Set Membership: |
|