Search
Search

CDK12| Gene Symbol: CDK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584019.2 | XP_005584076.1 | 7 - 8 | 3292 | 85 | |
| XM_005584020.2 | XP_005584077.1 | 7 - 8 | 3290 | 85 | ||
| XM_005584021.2 | XP_005584078.1 | 7 - 8 | 3293 | 85 | ||
| XM_015437898.1 | XP_015293384.1 | 8 - 9 | 2905 | 85 | ||
Set Membership: |
|