Search
Search

RNF111| Gene Symbol: RNF111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559668.2 | XP_005559725.1 | 8 - 9 | 2824 | 92 | |
| XM_005559669.2 | XP_005559726.1 | 8 - 9 | 2577 | 92 | ||
| XM_005559670.2 | XP_005559727.1 | 8 - 9 | 2709 | 92 | ||
| XM_005559671.2 | XP_005559728.1 | 8 - 9 | 2709 | 92 | ||
| XM_015452882.1 | XP_015308368.1 | 8 - 9 | 2823 | 92 | ||
Set Membership: |
|