Search
Search

CDHR5| Gene Symbol: CDHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576765.2 | XP_005576822.1 | 9 - 10 | 1267 | 171 | |
| XM_005576766.2 | XP_005576823.1 | 8 - 9 | 1439 | 171 | ||
| XM_005576767.2 | XP_005576824.1 | 9 - 10 | 1267 | 171 | ||
| XM_005576768.2 | XP_005576825.1 | 9 - 10 | 1267 | 171 | ||
| XM_015434220.1 | XP_015289706.1 | 9 - 10 | 1267 | 171 | ||
| XM_015434221.1 | XP_015289707.1 | 9 - 10 | 1267 | 171 | ||
Set Membership: |
|