Search
Search

TPCN1| Gene Symbol: TPCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572324.2 | XP_005572381.1 | 5 - 6 | 835 | 73 | |
| XM_005572325.2 | XP_005572382.1 | 5 - 6 | 761 | 73 | ||
| XM_015431589.1 | XP_015287075.1 | 4 - 5 | 1262 | 73 | ||
Set Membership: |
|