Search
Search

DZANK1| Gene Symbol: DZANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568255.2 | XP_005568312.1 | 3 - 4 | 911 | 87 | |
| XM_005568256.2 | XP_005568313.1 | 3 - 4 | 985 | 87 | ||
| XM_005568257.2 | XP_005568314.1 | 3 - 4 | 336 | 87 | ||
| XM_015457754.1 | XP_015313240.1 | 2 - 3 | 1271 | 87 | ||
| XM_015457755.1 | XP_015313241.1 | 2 - 3 | 1271 | 87 | ||
| XM_015457756.1 | XP_015313242.1 | 2 - 3 | 1271 | 87 | ||
| XM_015457757.1 | XP_015313243.1 | 2 - 3 | 1271 | 87 | ||
| XM_015457758.1 | XP_015313244.1 | 2 - 3 | 1271 | 87 | ||
Set Membership: |
|