Search
Search

SDK2| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584828.2 | XP_005584885.1 | 14 - 15 | 2855 | 100 | |
| XM_005584829.2 | XP_005584886.1 | 14 - 15 | 2855 | 100 | ||
Set Membership: |
|