Search
Search

UGGT2| Gene Symbol: UGGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439433.1 | XP_015294919.1 | 34 - 35 | 4055 | 94 | |
| XM_015439435.1 | XP_015294921.1 | 33 - 34 | 3947 | 94 | ||
| XM_015439436.1 | XP_015294922.1 | 32 - 33 | 3899 | 94 | ||
| XM_015439437.1 | XP_015294923.1 | 34 - 35 | 4055 | 94 | ||
| XM_015439438.1 | XP_015294924.1 | 34 - 35 | 4055 | 94 | ||
| XM_015439439.1 | XP_015294925.1 | 34 - 35 | 3746 | 94 | ||
| XM_015439440.1 | XP_015294926.1 | 29 - 30 | 3148 | 94 | ||
Set Membership: |
|