Search
Search

TXNDC16| Gene Symbol: TXNDC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561262.2 | XP_005561319.2 | 6 - 7 | 842 | 97 | |
| XM_015453789.1 | XP_015309275.1 | 6 - 7 | 886 | 97 | ||
| XM_015453790.1 | XP_015309276.1 | 6 - 7 | 886 | 97 | ||
Set Membership: |
|