Search
Search

KLHL8| Gene Symbol: KLHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555362.2 | XP_005555419.1 | 6 - 7 | 1560 | 142 | |
| XM_005555363.2 | XP_005555420.1 | 6 - 7 | 1633 | 142 | ||
| XM_005555364.2 | XP_005555421.1 | 6 - 7 | 1445 | 142 | ||
| XM_005555366.2 | XP_005555423.1 | 6 - 7 | 1543 | 142 | ||
| XM_015450635.1 | XP_015306121.1 | 6 - 7 | 2222 | 142 | ||
| XM_015450636.1 | XP_015306122.1 | 5 - 6 | 1014 | 142 | ||
Set Membership: |
|