Search
Search

FANCE| Gene Symbol: FANCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553219.2 | XP_005553276.1 | 2 - 3 | 905 | 155 | |
| XM_005553220.2 | XP_005553277.1 | 2 - 3 | 905 | 155 | ||
| XM_005553224.2 | XP_005553281.1 | 2 - 3 | 845 | 155 | ||
Set Membership: |
|