Search
Search

ACBD4| Gene Symbol: ACBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584489.2 | XP_005584546.1 | 9 - 10 | 1578 | 115 | |
| XM_005584490.2 | XP_005584547.1 | 7 - 8 | 1378 | 115 | ||
| XM_005584491.2 | XP_005584548.1 | 7 - 8 | 1374 | 115 | ||
| XM_015438627.1 | XP_015294113.1 | 9 - 10 | 1601 | 115 | ||
| XM_015438628.1 | XP_015294114.1 | 10 - 11 | 1507 | 115 | ||
| XM_015438630.1 | XP_015294116.1 | 10 - 11 | 1502 | 115 | ||
| XM_015438631.1 | XP_015294117.1 | 9 - 10 | 1583 | 115 | ||
| XM_015438632.1 | XP_015294118.1 | 7 - 8 | 1397 | 115 | ||
Set Membership: |
|