Search
Search

CPEB1| Gene Symbol: CPEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560321.2 | XP_005560378.1 | 8 - 9 | 1274 | 97 | |
| XM_005560323.2 | XP_005560380.1 | 7 - 8 | 1215 | 97 | ||
| XM_015453198.1 | XP_015308684.1 | 7 - 8 | 1184 | 97 | ||
Set Membership: |
|