Search
Search

SLC4A9| Gene Symbol: SLC4A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557958.2 | XP_005558015.1 | 7 - 8 | 1647 | 72 | |
| XM_005557959.2 | XP_005558016.1 | 7 - 8 | 1569 | 72 | ||
| XM_005557961.2 | XP_005558018.1 | 7 - 8 | 1569 | 72 | ||
| XM_005557969.2 | XP_005558026.1 | 6 - 7 | 719 | 72 | ||
| XM_015451963.1 | XP_015307449.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451964.1 | XP_015307450.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451965.1 | XP_015307451.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451966.1 | XP_015307452.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451967.1 | XP_015307453.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451968.1 | XP_015307454.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451970.1 | XP_015307456.1 | 6 - 7 | 1232 | 72 | ||
| XM_015451971.1 | XP_015307457.1 | 7 - 8 | 1569 | 72 | ||
Set Membership: |
|