Search
Search

FBXL13| Gene Symbol: FBXL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550406.2 | XP_005550463.2 | 12 - 13 | 1342 | 79 | |
| XM_005550410.2 | XP_005550467.1 | 11 - 12 | 1267 | 79 | ||
| XM_015447859.1 | XP_015303345.1 | 12 - 13 | 1342 | 79 | ||
| XM_015447860.1 | XP_015303346.1 | 12 - 13 | 1342 | 79 | ||
| XM_015447861.1 | XP_015303347.1 | 10 - 11 | 1154 | 79 | ||
| XM_015447862.1 | XP_015303348.1 | 12 - 13 | 1440 | 79 | ||
| XM_015447863.1 | XP_015303349.1 | 11 - 12 | 1240 | 79 | ||
| XM_015447864.1 | XP_015303350.1 | 12 - 13 | 1402 | 79 | ||
| XM_015447865.1 | XP_015303351.1 | 11 - 12 | 1372 | 79 | ||
| XM_015447866.1 | XP_015303352.1 | 12 - 13 | 1342 | 79 | ||
| GenBank mRNA | AB179462.1 | - | 11 - 12 | 1264 | 79 | |
Set Membership: |
|