Search
Search

HORMAD2| Gene Symbol: HORMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567640.2 | XP_005567697.1 | 5 - 6 | 415 | 137 | |
| XM_005567641.2 | XP_005567698.1 | 5 - 6 | 579 | 137 | ||
| XM_005567642.2 | XP_005567699.1 | 5 - 6 | 502 | 137 | ||
| XM_015457519.1 | XP_015313005.1 | 5 - 6 | 415 | 137 | ||
Set Membership: |
|