Search
Search

SYNE3| Gene Symbol: SYNE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562148.2 | XP_005562205.2 | 3 - 4 | 1248 | 143 | |
| XM_015454187.1 | XP_015309673.1 | 3 - 4 | 1249 | 143 | ||
Set Membership: |
|