Search
Search

EFCAB3| Gene Symbol: EFCAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584606.2 | XP_005584663.1 | 65 - 66 | 5099 | 117 | |
| XM_005584607.2 | XP_005584664.1 | 3 - 4 | 308 | 117 | ||
| XM_005584608.2 | XP_005584665.1 | 3 - 4 | 293 | 117 | ||
| XM_015438668.1 | XP_015294154.1 | 95 - 96 | 9317 | 117 | ||
| XM_015438669.1 | XP_015294155.1 | 96 - 97 | 8849 | 117 | ||
| XM_015438670.1 | XP_015294156.1 | 96 - 97 | 8782 | 117 | ||
| XM_015438671.1 | XP_015294157.1 | 93 - 94 | 9163 | 117 | ||
| XM_015438672.1 | XP_015294158.1 | 90 - 91 | 8894 | 117 | ||
| XM_015438673.1 | XP_015294159.1 | 95 - 96 | 9319 | 117 | ||
| XM_015438674.1 | XP_015294160.1 | 72 - 73 | 5759 | 117 | ||
| GenBank mRNA | AB074446.1 | - | 200 | 117 | ||
Set Membership: |
|