Search
Search

CNTN4| Gene Symbol: CNTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547640.2 | XP_005547697.1 | 5 - 6 | 615 | 141 | |
| XM_005547641.2 | XP_005547698.1 | 4 - 5 | 542 | 141 | ||
| XM_005547643.2 | XP_005547700.1 | 3 - 4 | 4235 | 141 | ||
| XM_015445393.1 | XP_015300879.1 | 5 - 6 | 627 | 141 | ||
| XM_015445394.1 | XP_015300880.1 | 3 - 4 | 3195 | 141 | ||
| XM_015445395.1 | XP_015300881.1 | 4 - 5 | 543 | 141 | ||
Set Membership: |
|