Search
Search

STARD13| Gene Symbol: STARD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585614.2 | XP_005585671.1 | 1 - 2 | 787 | 70 | |
| XM_015439138.1 | XP_015294624.1 | 1 - 2 | 787 | 70 | ||
Set Membership: |
|