Search
Search

TEX33| Gene Symbol: TEX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567393.2 | XP_005567450.1 | 4 - 5 | 748 | 111 | |
| XM_005567394.2 | XP_005567451.1 | 4 - 5 | 755 | 111 | ||
| GenBank mRNA | AB168671.1 | - | 4 - 5 | 742 | 111 | |
Set Membership: |
|