Search
Search

SFXN4| Gene Symbol: SFXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566561.2 | XP_005566618.1 | 6 - 7 | 420 | 114 | |
| XM_005566562.2 | XP_005566619.1 | 6 - 7 | 422 | 114 | ||
| XM_015456767.1 | XP_015312253.1 | 6 - 7 | 420 | 114 | ||
Set Membership: |
|