Search
Search

ADAMTS9| Gene Symbol: ADAMTS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547536.2 | XP_005547593.1 | 22 - 23 | 3691 | 71 | |
| XM_005547537.2 | XP_005547594.1 | 21 - 22 | 3291 | 71 | ||
| XM_005547538.2 | XP_005547595.1 | 21 - 22 | 3237 | 71 | ||
Set Membership: |
|