Search
Search

ADORA2A| Gene Symbol: ADORA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567785.2 | XP_005567842.1 | 2 - 3 | 1274 | 98 | |
| XM_005567786.2 | XP_005567843.1 | 2 - 3 | 706 | 98 | ||
| XM_015457572.1 | XP_015313058.1 | 2 - 3 | 1074 | 98 | ||
| XM_015457573.1 | XP_015313059.1 | 3 - 4 | 1391 | 98 | ||
| XM_015457574.1 | XP_015313060.1 | 2 - 3 | 932 | 98 | ||
| XM_015457575.1 | XP_015313061.1 | 2 - 3 | 662 | 98 | ||
| GenBank mRNA | AB172319.1 | - | 2 - 3 | 768 | 98 | |
Set Membership: |
|