Search
Search

HAS3| Gene Symbol: HAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592370.2 | XP_005592427.1 | 3 - 4 | 1609 | 61 | |
| XM_015443098.1 | XP_015298584.1 | 2 - 3 | 1240 | 61 | ||
| XM_015443099.1 | XP_015298585.1 | 2 - 3 | 929 | 61 | ||
Set Membership: |
|