Search
Search

RALGAPA1| Gene Symbol: RALGAPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561079.2 | XP_005561136.1 | 24 - 25 | 4158 | 129 | |
| XM_005561082.2 | XP_005561139.1 | 23 - 24 | 4017 | 129 | ||
| XM_005561084.2 | XP_005561141.1 | 24 - 25 | 3934 | 129 | ||
| XM_005561086.2 | XP_005561143.1 | 23 - 24 | 3793 | 129 | ||
| XM_005561087.2 | XP_005561144.1 | 23 - 24 | 3793 | 129 | ||
| XM_015453710.1 | XP_015309196.1 | 24 - 25 | 5535 | 129 | ||
| XM_015453711.1 | XP_015309197.1 | 24 - 25 | 5535 | 129 | ||
| XM_015453712.1 | XP_015309198.1 | 23 - 24 | 5394 | 129 | ||
| XM_015453713.1 | XP_015309199.1 | 24 - 25 | 5535 | 129 | ||
| XM_015453714.1 | XP_015309200.1 | 24 - 25 | 5535 | 129 | ||
| XM_015453715.1 | XP_015309201.1 | 19 - 20 | 4517 | 129 | ||
| XM_015453716.1 | XP_015309202.1 | 24 - 25 | 5535 | 129 | ||
Set Membership: |
|