Search
Search

ZYX| Gene Symbol: ZYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551043.2 | XP_005551100.2 | 5 - 6 | 1180 | 73 | |
| XM_005551044.2 | XP_005551101.1 | 5 - 6 | 1302 | 73 | ||
| XM_005551045.2 | XP_005551102.1 | 5 - 6 | 1142 | 73 | ||
Set Membership: |
|