Search
Search

MX1| Gene Symbol: MX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285184.1 | NP_001272113.1 | 4 - 5 | 581 | 72 | |
| XM_015446623.1 | XP_015302109.1 | 7 - 8 | 868 | 72 | ||
| XM_015446624.1 | XP_015302110.1 | 7 - 8 | 898 | 72 | ||
| XM_015446625.1 | XP_015302111.1 | 7 - 8 | 887 | 72 | ||
| GenBank mRNA | JX297237.1 | - | 4 - 5 | 581 | 72 | |
Set Membership: |
|