Search
Search

USP14| Gene Symbol: USP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285328.1 | NP_001272257.1 | 12 - 13 | 1194 | 108 | |
| XM_005587070.2 | XP_005587127.1 | 14 - 15 | 1537 | 108 | ||
| XM_005587071.2 | XP_005587128.1 | 13 - 14 | 1360 | 108 | ||
| GenBank mRNA | AB172341.1 | - | 12 - 13 | 1194 | 108 | |
Set Membership: |
|