Search
Search

NUCKS1| Gene Symbol: NUCKS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284904.1 | NP_001271833.1 | 5 - 6 | 441 | 102 | |
| XM_005540610.2 | XP_005540667.2 | 4 - 5 | 102 | |||
| GenBank mRNA | AB173740.1 | - | 5 - 6 | 441 | 102 | |
Set Membership: |
|