Search
Search

NADSYN1| Gene Symbol: NADSYN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283241.1 | NP_001270170.1 | 2 - 3 | 196 | 72 | |
| XM_005576939.2 | XP_005576996.1 | 2 - 3 | 339 | 72 | ||
| GenBank mRNA | AB169410.1 | - | 2 - 3 | 196 | 72 | |
Set Membership: |
|