Search
Search

PSMA4| Gene Symbol: PSMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283242.1 | NP_001270171.1 | 8 - 9 | 751 | 111 | |
| GenBank mRNA | AB168265.1 | - | 8 - 9 | 751 | 111 | |
| AB169649.1 | - | 8 - 9 | 768 | 111 | ||
Set Membership: |
|