Search
Search

SNCG| Gene Symbol: SNCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283870.1 | NP_001270799.1 | 2 - 3 | 246 | 83 | |
| XM_015455894.1 | XP_015311380.1 | 4 - 5 | 619 | 83 | ||
| GenBank mRNA | AB220471.1 | - | 2 - 3 | 246 | 83 | |
Set Membership: |
|