Search
Search

C6H5orf52| Gene Symbol: C6H5orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558397.2 | XP_005558454.2 | 1 - 2 | 478 | 116 | |
| XM_015452195.1 | XP_015307681.1 | 1 - 2 | 478 | 116 | ||
Set Membership: |
|