Search
Search

SLC28A1| Gene Symbol: SLC28A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560382.2 | XP_005560439.1 | 12 - 13 | 2521 | 58 | |
| XM_015453316.1 | XP_015308802.1 | 12 - 13 | 2549 | 58 | ||
| XM_015453317.1 | XP_015308803.1 | 11 - 12 | 2407 | 58 | ||
Set Membership: |
|