Search
Search

C16H17orf67| Gene Symbol: C16H17orf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583790.2 | XP_005583847.1 | 6 - 7 | 1080 | 82 | |
| XM_005583793.2 | XP_005583850.1 | 3 - 4 | 1042 | 82 | ||
| XM_015438324.1 | XP_015293810.1 | 7 - 8 | 1242 | 82 | ||
| XM_015438325.1 | XP_015293811.1 | 7 - 8 | 1757 | 82 | ||
| XM_015438326.1 | XP_015293812.1 | 7 - 8 | 1662 | 82 | ||
| XM_015438327.1 | XP_015293813.1 | 7 - 8 | 1644 | 82 | ||
Set Membership: |
|