Search
Search

GABRG3| Gene Symbol: GABRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559026.2 | XP_005559083.1 | 4 - 5 | 637 | 95 | |
| XM_015452551.1 | XP_015308037.1 | 4 - 5 | 638 | 95 | ||
| XM_015452552.1 | XP_015308038.1 | 2 - 3 | 825 | 95 | ||
Set Membership: |
|