Search
Search

SIGLEC14| Gene Symbol: SIGLEC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590135.2 | XP_005590192.1 | 6 - 7 | 1748 | 81 | |
| XM_005590136.2 | XP_005590193.1 | 9 - 10 | 3041 | 81 | ||
| XM_015441650.1 | XP_015297136.1 | 9 - 10 | 3041 | 81 | ||
| XM_015441651.1 | XP_015297137.1 | 8 - 9 | 3145 | 81 | ||
| XM_015441653.1 | XP_015297139.1 | 6 - 7 | 1510 | 81 | ||
Set Membership: |
|