Search
Search

LMNA| Gene Symbol: LMNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541485.2 | XP_005541542.1 | 6 - 7 | 1588 | 103 | |
| GenBank mRNA | AB125177.1 | - | 6 - 7 | 1365 | 103 | |
| AB220427.1 | - | 6 - 7 | 1365 | 103 | ||
Set Membership: |
|