Search
Search

BTBD17| Gene Symbol: BTBD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584838.2 | XP_005584895.1 | 3 - 4 | 1610 | 101 | |
| XM_015438800.1 | XP_015294286.1 | 3 - 4 | 1703 | 101 | ||
| XM_015438801.1 | XP_015294287.1 | 2 - 3 | 1546 | 101 | ||
Set Membership: |
|