Search
Search

PLIN4| Gene Symbol: PLIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587568.2 | XP_005587625.1 | 6 - 7 | 3742 | 113 | |
| XM_005587570.2 | XP_005587627.1 | 6 - 7 | 3759 | 113 | ||
| XM_005587571.2 | XP_005587628.1 | 4 - 5 | 3562 | 113 | ||
| XM_015440504.1 | XP_015295990.1 | 6 - 7 | 3760 | 113 | ||
| XM_015440506.1 | XP_015295992.1 | 6 - 7 | 3745 | 113 | ||
Set Membership: |
|